Learn More
Medchemexpress LLC Dimethyl adipate | 627-93-0 | MFCD00008469 | >99.0% | 174.19 g/mol | C8H14O4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW015301R100MG
Encompass_Preferred
Dimethyl adipate is the dimethyl ester of adipic (hexanedioic) acid (C8H14O4, MW 174.19 g/mol) used as an analytical reference standard for research and analytical applications. It is an aliphatic dicarboxylate commonly employed as a plasticizer, solvent, and synthetic intermediate.
  • High purity reference material for analytical assays.
  • Suitable as a solvent and plasticizer in polymer studies.
  • Stable aliphatic dicarboxylate with predictable physical properties.
  • Molecular formula C8H14O4 and molecular weight 174.19 g/mol.
  • Available in small reference quantities for laboratory use.
Catalog No. 50-003-03707