Learn More
Medchemexpress LLC Dimethyl adipate | 627-93-0 | MFCD00008469 | 99.0% | 174.19 g·mol⁻¹ | C8H14O4 | 250 MG
SDP
Catalog No. 5000302780
Encompass_Preferred
Dimethyl adipate is the analytical reference standard of hexanedioic acid dimethyl ester used for qualitative and quantitative analysis by HPLC, GC, and mass spectrometry. It is supplied with verified identity data (1H NMR and MS) and controlled impurity levels for use in research, method development, and assay calibration. Store refrigerated, sealed, and protected from light and moisture.
  • Intended for analytical HPLC, GC, and mass spectrometry applications.
  • High purity: HPLC purity 99.01% (assay 98.70%).
  • Identity confirmed by 1H NMR and MS.
  • Low residue on ignition and low moisture content.
  • Supplied as a colorless liquid in a brown glass bottle.
  • Storage: 4°C, sealed, protect from light and moisture.
Catalog No. 50-003-02780 Supplier Medchemexpress LLC Supplier No. 250MGHYW015301R
Dimethyl adipate is the analytical reference standard of hexanedioic acid dimethyl ester used for qualitative and quantitative analysis by HPLC, GC, and mass spectrometry. It is supplied with verified identity data (1H NMR and MS) and controlled impurity levels for use in research, method development, and assay calibration. Store refrigerated, sealed, and protected from light and moisture.
  • Intended for analytical HPLC, GC, and mass spectrometry applications.
  • High purity: HPLC purity 99.01% (assay 98.70%).
  • Identity confirmed by 1H NMR and MS.
  • Low residue on ignition and low moisture content.
  • Supplied as a colorless liquid in a brown glass bottle.
  • Storage: 4°C, sealed, protect from light and moisture.