Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dimethyl adipate-d4-1 | 55724-08-8 | 99.4% | 178.22 | C8H10D4O4 | 100 MG
SDP

Catalog No. 5000302963
Encompass_Preferred

Dimethyl adipate-d4-1 is the deuterium-labeled analogue of dimethyl adipate supplied as a stable isotope reference standard for analytical and tracer applications. It is provided as a high-purity material intended for mass spectrometry, quantitative analysis, and method development.

  • Deuterium-labeled standard for mass spectrometry and tracer studies.
  • High purity, 99.4%.
  • Chemical formula C8H10D4O4; molecular weight 178.22.
  • Packaged as a 100 mg vial for analytical use.
  • Accompanied by certificate of analysis and safety data sheet.

Catalog No. 50-003-02963 Supplier Medchemexpress LLC Supplier No. HYW015301S1100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dimethyl adipate-d4-1 is the deuterium-labeled analogue of dimethyl adipate supplied as a stable isotope reference standard for analytical and tracer applications. It is provided as a high-purity material intended for mass spectrometry, quantitative analysis, and method development.

  • Deuterium-labeled standard for mass spectrometry and tracer studies.
  • High purity, 99.4%.
  • Chemical formula C8H10D4O4; molecular weight 178.22.
  • Packaged as a 100 mg vial for analytical use.
  • Accompanied by certificate of analysis and safety data sheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.