Learn More
Medchemexpress LLC Dimethyl adipate-d4 (hexanedioic acid dimethyl ester-d4) | 55724-08-8 | 99.4% | 178.22 g/mol | C8H10D4O4 | 10 MG
SDP
Catalog No. 5000302929
Encompass_Preferred
Dimethyl adipate-d4-1 is the deuterium-labeled analogue of dimethyl adipate (hexanedioic acid dimethyl ester) provided as an analytical standard for use as a tracer or internal standard in quantitative techniques such as NMR, GC-MS, and LC-MS. Supplied in small milligram quantities for research use, it supports accurate mass spectrometric and spectroscopic analysis.
Catalog No. 50-003-02929 Supplier Medchemexpress LLC Supplier No. HYW015301S110MG
Dimethyl adipate-d4-1 is the deuterium-labeled analogue of dimethyl adipate (hexanedioic acid dimethyl ester) provided as an analytical standard for use as a tracer or internal standard in quantitative techniques such as NMR, GC-MS, and LC-MS. Supplied in small milligram quantities for research use, it supports accurate mass spectrometric and spectroscopic analysis.