Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dimethyl adipate-d4 (hexanedioic acid dimethyl ester-d4) | 55724-08-8 | 99.4% | 178.22 g·mol⁻¹ | C8H10D4O4 | 50 MG
SDP

Catalog No. 5000302784
Encompass_Preferred

Dimethyl adipate-d4 is a deuterium-labeled analog of dimethyl adipate supplied as a high-purity isotopic standard for analytical and research use. It is intended for use in mass spectrometry, quantitative analysis, and synthetic applications where a deuterated reference is required. Supplied as a small-scale research vial for laboratory use only.

  • High isotopic labeling for reliable mass spectrometric quantitation.
  • Purity 99.4% (GC) for consistent analytical performance.
  • Molecular weight 178.22 g·mol⁻¹ for accurate calculations.
  • Chemical formula C8H10D4O4 indicates deuterium substitution.
  • Stable liquid form suitable for standard preparation and spiking.
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-003-02784 Supplier Medchemexpress LLC Supplier No. HYW015301S150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dimethyl adipate-d4 is a deuterium-labeled analog of dimethyl adipate supplied as a high-purity isotopic standard for analytical and research use. It is intended for use in mass spectrometry, quantitative analysis, and synthetic applications where a deuterated reference is required. Supplied as a small-scale research vial for laboratory use only.

  • High isotopic labeling for reliable mass spectrometric quantitation.
  • Purity 99.4% (GC) for consistent analytical performance.
  • Molecular weight 178.22 g·mol⁻¹ for accurate calculations.
  • Chemical formula C8H10D4O4 indicates deuterium substitution.
  • Stable liquid form suitable for standard preparation and spiking.
  • Intended for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.