Learn More
Medchemexpress LLC Dimethyl adipate-d8 | 52089-64-2 | MFCD12022192 | 98.0% | 182.24 | C8H6D8O4 | 100 MG
SDP
Catalog No. 5000303738
Encompass_Preferred
Dimethyl adipate-d8 is a deuterium-labeled analogue of dimethyl adipate used as an isotopically labeled reference standard for analytical chemistry. It is intended for use as an internal standard in mass spectrometry and chromatographic assays, offering predictable chromatographic behavior with a distinct mass shift.
  • Deuterated analogue with eight deuterium atoms, providing clear mass separation.
  • High isotopic enrichment for reliable internal standard performance.
  • High chemical purity suitable for analytical workflows.
  • Multiple small package sizes available for laboratory-scale use.
  • Stable under recommended storage conditions for consistent results.
Catalog No. 50-003-03738 Supplier Medchemexpress LLC Supplier No. HYW015301S2100MG
Dimethyl adipate-d8 is a deuterium-labeled analogue of dimethyl adipate used as an isotopically labeled reference standard for analytical chemistry. It is intended for use as an internal standard in mass spectrometry and chromatographic assays, offering predictable chromatographic behavior with a distinct mass shift.
  • Deuterated analogue with eight deuterium atoms, providing clear mass separation.
  • High isotopic enrichment for reliable internal standard performance.
  • High chemical purity suitable for analytical workflows.
  • Multiple small package sizes available for laboratory-scale use.
  • Stable under recommended storage conditions for consistent results.