Learn More
Medchemexpress LLC Dimethyl adipate-d8 | 52089-64-2 | MFCD12022192 | 98.0% | 182.24 g/mol | C8H6D8O4 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW015301S21MG
Encompass_Preferred
Dimethyl adipate-d8 is the deuterium-labeled analog of dimethyl adipate (hexanedioic acid dimethyl ester), provided as an isotopically labeled reference standard for analytical and mass spectrometry workflows. It is supplied at high purity for use as an internal standard, tracer, or calibration compound in quantitative assays and method development.
Catalog No. 50-003-03749