Learn More
Medchemexpress LLC Dimethyl adipate (hexanedioic acid dimethyl ester) | 627-93-0 | MFCD00008469 | 100.0% | 174.19 g/mol | C8H14O4 | 100 G
SDP
Catalog No. 5000302925
Encompass_Preferred
Dimethyl adipate (hexanedioic acid dimethyl ester) is an aliphatic dicarboxylate ester used as a plasticizer and as an intermediate in organic synthesis. Supplied as a high-purity liquid for research and analytical applications, it is documented with a datasheet, COA, and SDS, and is stable when stored under recommended conditions.
  • High purity (99.98%) suitable for analytical use.
  • Colorless to light yellow liquid; density 1.06 g/cm3.
  • Functions as a plasticizer and synthetic intermediate.
  • Available in multiple pack sizes, including 100 g.
  • Documented quality with datasheet, COA, and SDS available.
  • Stable under recommended storage to support long shelf life.
Catalog No. 50-003-02925 Supplier Medchemexpress LLC Supplier No. HYW015301100G
Dimethyl adipate (hexanedioic acid dimethyl ester) is an aliphatic dicarboxylate ester used as a plasticizer and as an intermediate in organic synthesis. Supplied as a high-purity liquid for research and analytical applications, it is documented with a datasheet, COA, and SDS, and is stable when stored under recommended conditions.
  • High purity (99.98%) suitable for analytical use.
  • Colorless to light yellow liquid; density 1.06 g/cm3.
  • Functions as a plasticizer and synthetic intermediate.
  • Available in multiple pack sizes, including 100 g.
  • Documented quality with datasheet, COA, and SDS available.
  • Stable under recommended storage to support long shelf life.