Learn More
Medchemexpress LLC Dimethyl adipate (hexanedioic acid dimethyl ester) | 627-93-0 | MFCD00008469 | 100.0% | 174.19 g/mol | C8H14O4 | 50 G
SDP
Catalog No. 5000303642
Encompass_Preferred
Dimethyl adipate (hexanedioic acid dimethyl ester) is an aliphatic dicarboxylate ester used as a plasticizer, an intermediate in organic synthesis, and as an analytical standard for research applications. The compound has the molecular formula C8H14O4 and a molecular weight of 174.19 g/mol.
  • High purity suitable for analytical and research use.
  • Functions as a plasticizer and synthetic intermediate.
  • Molecular formula C8H14O4 and molecular weight 174.19 g/mol.
  • Commonly supplied in laboratory pack sizes including 50 g.
  • Stable when stored under recommended conditions for analytical standards.
Catalog No. 50-003-03642 Supplier Medchemexpress LLC Supplier No. HYW01530150G
Dimethyl adipate (hexanedioic acid dimethyl ester) is an aliphatic dicarboxylate ester used as a plasticizer, an intermediate in organic synthesis, and as an analytical standard for research applications. The compound has the molecular formula C8H14O4 and a molecular weight of 174.19 g/mol.
  • High purity suitable for analytical and research use.
  • Functions as a plasticizer and synthetic intermediate.
  • Molecular formula C8H14O4 and molecular weight 174.19 g/mol.
  • Commonly supplied in laboratory pack sizes including 50 g.
  • Stable when stored under recommended conditions for analytical standards.