Learn More
Cayman Chemical Dimethyl biphenyl-4,4'-dicarboxylate, Purity: ≥95%, CAS: 792-74-5, 100 g
SDP
Catalog No. 502967384
Encompass_Preferred
A planar biphenyldicarboxylic acid ester used as a hepatoprotectant in the treatment of chronic hepatitis; 250 µg/ml stimulates JAK/STAT signaling and induces the expression of interferon-α stimulated genes in a HepG2 cell line
Catalog No. 50-296-7384 Supplier Cayman Chemical Supplier No. 17869100 G
A planar biphenyldicarboxylic acid ester used as a hepatoprotectant in the treatment of chronic hepatitis; 250 µg/ml stimulates JAK/STAT signaling and induces the expression of interferon-α stimulated genes in a HepG2 cell line