Learn More
Medchemexpress LLC Dimethyl biphenyl-4,4'-dicarboxylate | 792-74-5 | 99.9% | 1 G
SDP
Catalog No. 5000339574
Encompass_Preferred
Dimethyl biphenyl-4,4'-dicarboxylate is a hepatoprotective agent that can be obtained from Schizandra fructus. It functions by stimulating the Jak/Stat signaling pathway, inducing IFN-α-stimulated genes like 6-16 and ISG12, and inhibiting pregenomic RNA and HBeAg replication. It is used as an auxiliary improving agent for chronic hepatitis and applicable for research related to chronic hepatitis B.
  • Functions as a hepatoprotective agent.
  • Stimulates the Jak/Stat signaling pathway.
  • Induces expression of IFN-α-stimulated genes (6-16, ISG12).
  • Inhibits replication of pregenomic RNA and HBeAg.
  • Can be loaded into polymer micelles for delivery.
  • Supports research on chronic hepatitis B.
  • Induces expression of antiviral effector genes PKR and OAS.
Catalog No. 50-003-39574 Supplier Medchemexpress LLC Supplier No. HY1288541G
Dimethyl biphenyl-4,4'-dicarboxylate is a hepatoprotective agent that can be obtained from Schizandra fructus. It functions by stimulating the Jak/Stat signaling pathway, inducing IFN-α-stimulated genes like 6-16 and ISG12, and inhibiting pregenomic RNA and HBeAg replication. It is used as an auxiliary improving agent for chronic hepatitis and applicable for research related to chronic hepatitis B.
  • Functions as a hepatoprotective agent.
  • Stimulates the Jak/Stat signaling pathway.
  • Induces expression of IFN-α-stimulated genes (6-16, ISG12).
  • Inhibits replication of pregenomic RNA and HBeAg.
  • Can be loaded into polymer micelles for delivery.
  • Supports research on chronic hepatitis B.
  • Induces expression of antiviral effector genes PKR and OAS.