Learn More
Enzo Life Sciences Dimethyl-bisphenol A (50mg). CAS: 1568-83-8
SDP
Supplier:  Enzo Life Sciences BMLGR3390050
Encompass_Preferred
A HIF-1α degradation promotor. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (>25 mg/ml) or ethanol (>25 mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0993