Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Bromomalonate 95.0+%, TCI America™
SDP

Catalog No. D388725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D388725G 25 g
D38875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D388725G Supplier TCI America Supplier No. D388725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 868-26-8
Molecular Formula C5H7BrO4
Molecular Weight (g/mol) 211.011
MDL Number MFCD00025865
InChI Key NEMOJKROKMMQBQ-UHFFFAOYSA-N
Synonym dimethyl bromomalonate, dimethyl 2-bromomalonate, dimethylbromomalonate, propanedioic acid, bromo-, dimethyl ester, 2-bromo dimethyl malonate, 1,3-dimethyl 2-bromopropanedioate, dimethylbromomalo-nate, acmc-209qbk, dimethyl-2-bromomalonate, dimethyl 2-bromomalonate #
PubChem CID 70087
IUPAC Name dimethyl 2-bromopropanedioate
SMILES COC(=O)C(C(=O)OC)Br

Specifications

Color Yellow
Boiling Point 225°C
Quantity 25 g
UN Number 3265
Formula Weight 211.01
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl Bromomalonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.