Learn More
Sigma Aldrich Fine Chemicals Biosciences Dimethyl carbonate Reagent500G
Catalog No. 502898953
Encompass
Dimethyl carbonate (DMC) is a versatile non-toxic biodegradable reagent with tunable chemical reactivity. It serves as a green alternative to dimethyl sulfate or methyl halides and phosgene for methylation and carboxylation reactions. The base-catalyzed reaction between carbon dioxide and methanol to form DMC has been studied. Various thermodynamic parameters of DMC have been reported.We are committed to bringing you Greener Alternative Products which adhere to one or more of The 12 Principles of Greener Chemistry. This product is a Greener alternative to conventional solvents and chemicals. Click here for more information.
Catalog No. 50-289-8953 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D152927500G
Dimethyl carbonate (DMC) is a versatile non-toxic biodegradable reagent with tunable chemical reactivity. It serves as a green alternative to dimethyl sulfate or methyl halides and phosgene for methylation and carboxylation reactions. The base-catalyzed reaction between carbon dioxide and methanol to form DMC has been studied. Various thermodynamic parameters of DMC have been reported.We are committed to bringing you Greener Alternative Products which adhere to one or more of The 12 Principles of Greener Chemistry. This product is a Greener alternative to conventional solvents and chemicals. Click here for more information.