Learn More
Medchemexpress LLC Dimethyl Citric acid | 53798-97-3 | ≥97.0% | 220.18 | 100 MG
SDP
Catalog No. 5000426102
Encompass_Preferred
Dimethyl Citric acid is a highly efficient deep eutectic solvent (DES) that serves as both a catalyst and a green reaction medium. It is suitable for the synthesis of various compounds, including bis(indolyl)methanes, quinolines, and aryl-4, 5-diphenyl-1H-imidazoles.
  • Highly efficient deep eutectic solvent (DES)
  • Functions as a catalyst
  • Acts as a green reaction medium
  • Used for synthesizing bis(indolyl)methanes
  • Used for synthesizing quinolines
  • Used for synthesizing aryl-4, 5-diphenyl-1H-imidazoles
Catalog No. 50-004-26102 Supplier Medchemexpress LLC Supplier No. HYN9542100MG
Dimethyl Citric acid is a highly efficient deep eutectic solvent (DES) that serves as both a catalyst and a green reaction medium. It is suitable for the synthesis of various compounds, including bis(indolyl)methanes, quinolines, and aryl-4, 5-diphenyl-1H-imidazoles.
  • Highly efficient deep eutectic solvent (DES)
  • Functions as a catalyst
  • Acts as a green reaction medium
  • Used for synthesizing bis(indolyl)methanes
  • Used for synthesizing quinolines
  • Used for synthesizing aryl-4, 5-diphenyl-1H-imidazoles