Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dimethyl cyclohexane-1,4-dicarboxylate, mixture of cis and trans

Catalog No. 20643125G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-643-125G 25 g
20-643-1100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 20-643-125G Supplier Sigma Aldrich Supplier No. 20643125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dimethyl cyclohexane-1,4-dicarboxylate, mixture of cis and trans may be used in chemical synthesis studies.

Specifications

CAS 94-60-0
Density 1.111 g/mL (at 25°C)
Quantity 25 g
Refractive Index n20/D 1.458 (literature)
Molecular Formula C10H16O4
Linear Formula C6H10(CO2CH3)2
MDL Number MFCD00001460
Boiling Point 131°C to 133°C (11 mmHg)
Molecular Weight (g/mol) 200.23
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.