Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl disulfide, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019678 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9678 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9678 Supplier MilliporeSigma Supelco Supplier No. 689861ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dimethyl disulfide is an organic disulfide, and a commonly used potent odorant which contributes to the aroma in heated food products.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 624-92-0
Ignition Point >572°F
Vapor Pressure 22 mmHg (20°C)
Vapor Density 3.24 (vs. air)
Density 1.046 g/mL (at 25°C)
Flash Point 15°C - closed cup
Physical Form Neat
Linear Formula CH3 SSCH3
MDL Number MFCD00008561
Melting Point -85°C
Boiling Point 109°C
Synonym DMDS; Methyl disulfide
Type Analytical Standard
Quantity 1 mL
Formula Weight 94.20
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.