Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Esculetin, MP Biomedicals

Catalog No. ICN222356 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN222356 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN222356 Supplier MP Biomedicals, Inc Supplier No. 0522235680

Chemical Identifiers

CAS 120-08-1
Molecular Formula C11H10O4
Molecular Weight (g/mol) 206.20
MDL Number MFCD00006871
InChI Key GUAFOGOEJLSQBT-UHFFFAOYSA-N
Synonym scoparone, 6,7-dimethoxycoumarin, 6,7-dimethylesculetin, escoparone, scoparon, aesculetin dimethyl ether, esculetin dimethyl ether, 6,7-dimethoxy-2-benzopyrone, scopoletin methyl ether, 6,7-dimethoxybenzopyran-2-one
PubChem CID 8417
ChEBI CHEBI:9055
IUPAC Name 6,7-dimethoxy-2H-chromen-2-one
SMILES COC1=C(OC)C=C2C=CC(=O)OC2=C1

Specifications

Quantity 100 mg
Formula Weight 206.21
Chemical Name or Material Dimethyl Esculetin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.