Learn More
Supelco Inc Dimethyl ether, 115-10-6, MFCD00008494, 100 G
Catalog No. NC0540432
Encompass
Linear Formula: (CH3)2O, Molecular Weight: 46.07, Synonym: DME, Methyl ether, Assay: ≥99%, BP: −24.8 °C(lit.), MP: −141 °C (lit.). Dimethyl ether (DME) can be used as a precursor to synthesize methyl acetate, oligomeric oxymethylene ethers (OMEs), polyoxymethylene dimethyl ethers (DMMx) and light olefins.
Catalog No. NC0540432 Supplier Supelco Inc Supplier No. 295299100G
Linear Formula: (CH3)2O, Molecular Weight: 46.07, Synonym: DME, Methyl ether, Assay: ≥99%, BP: −24.8 °C(lit.), MP: −141 °C (lit.). Dimethyl ether (DME) can be used as a precursor to synthesize methyl acetate, oligomeric oxymethylene ethers (OMEs), polyoxymethylene dimethyl ethers (DMMx) and light olefins.