Learn More
Ambeed DIMETHYL FUMARATE-100G
SDP
Catalog No. NC2161818
Encompass_Preferred
Dimethyl fumarate, 100G, CAS# 624-49-7, MFCD00064438, MOLWT: 144.13
This item is not returnable. View return policy
Catalog No. NC2161818 Supplier Ambeed Supplier No. A258585100G
Dimethyl fumarate, 100G, CAS# 624-49-7, MFCD00064438, MOLWT: 144.13