Learn More
Selleck Chemical LLC Dimethyl Fumarate-500mg
SDP
Catalog No. 502390002
Encompass_Preferred
Dimethyl fumarate (DMF,Tecfidera,BG-12) is the methyl ester of fumaric acid, used to treat people with relapsing forms of multiple sclerosis. Dimethyl fumarate is a nuclear factor (erythroid-derived)-like 2 (Nrf2) pathway activator and induces upregulation of antioxidant gene expression.
Catalog No. 50-239-0002 Supplier Selleck Chemical LLC Supplier No. S2586500MG
Dimethyl fumarate (DMF,Tecfidera,BG-12) is the methyl ester of fumaric acid, used to treat people with relapsing forms of multiple sclerosis. Dimethyl fumarate is a nuclear factor (erythroid-derived)-like 2 (Nrf2) pathway activator and induces upregulation of antioxidant gene expression.