Learn More
Medchemexpress LLC HY-17363 500mg Medchemexpress, Dimethyl fumarate CAS:624-49-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY17363500MG
Encompass_Preferred
Medchemexpress, HY-17363 500mg Dimethyl fumarate CAS:624-49-7 Dimethyl fumarate is a nuclear factor (erythroid-derived)-like 2 (Nrf2) pathway activator and induces upregulation of antioxidant gene expression. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0762