Learn More
Cayman Chemical Dimethyl fumarate, Purity: ≥95%, CAS: 624-49-7, 50 g
SDP
Catalog No. 502969489
Encompass_Preferred
A lipophilic electrophile that has profound effects on oxidative and immunological pathways; activates the transcription factor Nrf2, resulting in heme oxygenase-1 expression and increased glutathione levels; suppresses the expression of pro-inflammatory cytokines and inhibits angiogenesis
Catalog No. 50-296-9489 Supplier Cayman Chemical Supplier No. 1471450 G
A lipophilic electrophile that has profound effects on oxidative and immunological pathways; activates the transcription factor Nrf2, resulting in heme oxygenase-1 expression and increased glutathione levels; suppresses the expression of pro-inflammatory cytokines and inhibits angiogenesis