Learn More
Medchemexpress LLC Dimethyl fumarate | 624-49-7 | 100.0% | 144.13 | 1 ML
SDP
Catalog No. 5000385992
Encompass_Preferred
Dimethyl fumarate (DMF) is an orally active and brain-penetrant Nrf2 activator that induces upregulation of antioxidant gene expression. It also induces necroptosis in colon cancer cells via the GSH depletion/ROS increase/MAPKs activation pathway, and promotes cell autophagy. Dimethyl fumarate is used for multiple sclerosis research.
  • Orally active and brain-penetrant Nrf2 activator
  • Induces upregulation of antioxidant gene expression
  • Induces necroptosis in colon cancer cells through GSH depletion/ROS increase/MAPKs activation pathway
  • Promotes cell autophagy
  • Used for multiple sclerosis research
Catalog No. 50-003-85992 Supplier Medchemexpress LLC Supplier No. HY1736310MM/1ML
Dimethyl fumarate (DMF) is an orally active and brain-penetrant Nrf2 activator that induces upregulation of antioxidant gene expression. It also induces necroptosis in colon cancer cells via the GSH depletion/ROS increase/MAPKs activation pathway, and promotes cell autophagy. Dimethyl fumarate is used for multiple sclerosis research.
  • Orally active and brain-penetrant Nrf2 activator
  • Induces upregulation of antioxidant gene expression
  • Induces necroptosis in colon cancer cells through GSH depletion/ROS increase/MAPKs activation pathway
  • Promotes cell autophagy
  • Used for multiple sclerosis research