Learn More
Medchemexpress LLC Dimethyl fumarate | 624-49-7 | 100.0% | 144.13 | 25 MG
SDP
Catalog No. 5000384322
Encompass_Preferred
Dimethyl fumarate (DMF) is an orally active and brain-penetrant Nrf2 activator that induces upregulation of antioxidant gene expression. It induces necroptosis in colon cancer cells through GSH depletion/ROS increase/MAPKs activation pathway and also induces cell autophagy. Dimethyl fumarate can be used for multiple sclerosis research. It also acts as an immune modulator and inducer of the antioxidant response, suppressing HIV replication and neurotoxin release.
  • Orally active and brain-penetrant Nrf2 activator
  • Induces upregulation of antioxidant gene expression
  • Induces necroptosis in colon cancer cells
  • Induces cell autophagy
  • Used for multiple sclerosis research
  • Immune modulator
  • Inducer of the antioxidant response
  • Suppresses HIV replication and neurotoxin release
  • Inhibits dendritic cell (DC) maturation
  • Impairs NF-κB signaling
Catalog No. 50-003-84322 Supplier Medchemexpress LLC Supplier No. 25MGHY17363R
Dimethyl fumarate (DMF) is an orally active and brain-penetrant Nrf2 activator that induces upregulation of antioxidant gene expression. It induces necroptosis in colon cancer cells through GSH depletion/ROS increase/MAPKs activation pathway and also induces cell autophagy. Dimethyl fumarate can be used for multiple sclerosis research. It also acts as an immune modulator and inducer of the antioxidant response, suppressing HIV replication and neurotoxin release.
  • Orally active and brain-penetrant Nrf2 activator
  • Induces upregulation of antioxidant gene expression
  • Induces necroptosis in colon cancer cells
  • Induces cell autophagy
  • Used for multiple sclerosis research
  • Immune modulator
  • Inducer of the antioxidant response
  • Suppresses HIV replication and neurotoxin release
  • Inhibits dendritic cell (DC) maturation
  • Impairs NF-κB signaling