Learn More
Apexbio Technology LLC Dimethyl Fumarate 624-49-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000090233
Encompass_Preferred
Dimethyl fumarate (CAS 624-49-7) is a small-molecule activator targeting the nuclear factor erythroid 2-related factor 2 (Nrf2) signaling pathway It is designed to promote Nrf2 nuclear translocation and induce transcription of antioxidant response genes thereby modulating oxidative stress response pathways Dimethyl fumarate exerts its biological activity primarily through activation of Nrf2-mediated antioxidant gene expression In cell-based assays dimethyl fumarate demonstrates Nrf2 activation activity with an IC50 value of approximately 5 M Based on these pharmacological properties dimethyl fumarate holds research potential in the study of oxidative stress inflammation and related pathophysiological processes
Catalog No. 50-000-90233 Supplier Apexbio Technology LLC Supplier No. B19315.1
Dimethyl fumarate (CAS 624-49-7) is a small-molecule activator targeting the nuclear factor erythroid 2-related factor 2 (Nrf2) signaling pathway It is designed to promote Nrf2 nuclear translocation and induce transcription of antioxidant response genes thereby modulating oxidative stress response pathways Dimethyl fumarate exerts its biological activity primarily through activation of Nrf2-mediated antioxidant gene expression In cell-based assays dimethyl fumarate demonstrates Nrf2 activation activity with an IC50 value of approximately 5 M Based on these pharmacological properties dimethyl fumarate holds research potential in the study of oxidative stress inflammation and related pathophysiological processes