Learn More
Medchemexpress LLC Dimethyl fumarate | 624-49-7 | MFCD00064438 | 99.9% | 144.13 | C6H8O4 | 10 MG
SDP
Catalog No. 5000295430
Encompass_Preferred
Dimethyl fumarate is an analytical reference standard supplied as a white to off-white solid with high purity for research and analytical applications. It is commonly used in studies of Nrf2 activation, oxidative stress pathways, and multiple sclerosis research.
  • High purity (99.85%).
  • Solid, white to off-white physical form.
  • Chemical formula C6H8O4 and molecular weight 144.13 g/mol.
  • Available in small package sizes, including 10 mg.
  • Intended for research and analytical use in pathway and disease studies.
Catalog No. 50-002-95430 Supplier Medchemexpress LLC Supplier No. HY17363R10MG
Dimethyl fumarate is an analytical reference standard supplied as a white to off-white solid with high purity for research and analytical applications. It is commonly used in studies of Nrf2 activation, oxidative stress pathways, and multiple sclerosis research.
  • High purity (99.85%).
  • Solid, white to off-white physical form.
  • Chemical formula C6H8O4 and molecular weight 144.13 g/mol.
  • Available in small package sizes, including 10 mg.
  • Intended for research and analytical use in pathway and disease studies.