Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl fumarate, 99%

Catalog No. AC222181000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC222181000 100 g Glass bottle
AC222180250 25 g Glass bottle
AC222185000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC222181000 Supplier Thermo Scientific Chemicals Supplier No. 222181000
Add to Cart

CAS: 624-49-7 | C6H8O4 | 144.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-49-7
Molecular Formula C6H8O4
Molecular Weight (g/mol) 144.13
MDL Number MFCD00064438
InChI Key LDCRTTXIJACKKU-ONEGZZNKSA-N
Synonym dimethyl fumarate, tecfidera, e-dimethyl fumarate, fumaderm, dimethyl e-but-2-enedioate, fumaric acid, dimethyl ester, boletic acid dimethyl ester, fumaric acid dimethyl ester, methyl fumarate, dimethyl trans-ethylenedicarboxylate
PubChem CID 637568
ChEBI CHEBI:76004
IUPAC Name dimethyl (E)-but-2-enedioate
SMILES COC(=O)C=CC(=O)OC

Specifications

Melting Point 102°C to 105°C
Color Colorless to Yellow
Boiling Point 192°C to 193°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3O2CCH=CHCO2CH3
Quantity 100 g
Beilstein 02, 741
Solubility Information Solubility in water: 1.6g/L (20°C)
Formula Weight 144.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Dimethyl fumarate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/e
EINECSNumber 210-849-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.