Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dimethyl fumarate-d2 | 23057-98-9 | MFCD08704934 | 99.2% | 146.14 g/mol | C6H6D2O4 | 10 MG
SDP

Catalog No. 5000295431
Encompass_Preferred

Dimethyl fumarate-d2 is the deuterium-labeled isotopologue of dimethyl fumarate provided as a research reference standard. It is used in analytical, pharmacokinetic, and mechanistic studies where a mass-shifted tracer or internal standard is required, enabling precise detection and quantitation by mass spectrometry.

  • Deuterium-labeled for mass spectrometry and tracer studies.
  • Suitable as an internal standard in analytical and pharmacokinetic assays.
  • High chemical purity: 99.2%.
  • Available in small research quantities (10 mg) for analytical workflows.
  • Molecular formula C6H6D2O4; molecular weight 146.14 g/mol.

Catalog No. 50-002-95431 Supplier Medchemexpress LLC Supplier No. HY17363S110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dimethyl fumarate-d2 is the deuterium-labeled isotopologue of dimethyl fumarate provided as a research reference standard. It is used in analytical, pharmacokinetic, and mechanistic studies where a mass-shifted tracer or internal standard is required, enabling precise detection and quantitation by mass spectrometry.

  • Deuterium-labeled for mass spectrometry and tracer studies.
  • Suitable as an internal standard in analytical and pharmacokinetic assays.
  • High chemical purity: 99.2%.
  • Available in small research quantities (10 mg) for analytical workflows.
  • Molecular formula C6H6D2O4; molecular weight 146.14 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.