Learn More
Medchemexpress LLC Dimethyl fumarate-d6 | 66487-95-4 | 99.0% | 150.16 | 5 MG
SDP
Catalog No. 5000385486
Encompass_Preferred
Dimethyl fumarate-d6 is a deuterium labeled Dimethyl fumarate. Dimethyl fumarate is a nuclear factor (erythroid-derived)-like 2 (Nrf2) pathway activator and induces upregulation of antioxidant gene expression. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Used as a tracer and internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Activates the nuclear factor (erythroid-derived)-like 2 (Nrf2) pathway.
  • Upregulates antioxidant gene expression.
Catalog No. 50-003-85486 Supplier Medchemexpress LLC Supplier No. HY17363S5MG
Dimethyl fumarate-d6 is a deuterium labeled Dimethyl fumarate. Dimethyl fumarate is a nuclear factor (erythroid-derived)-like 2 (Nrf2) pathway activator and induces upregulation of antioxidant gene expression. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Used as a tracer and internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Activates the nuclear factor (erythroid-derived)-like 2 (Nrf2) pathway.
  • Upregulates antioxidant gene expression.