Learn More
Medchemexpress LLC Dimethyl fumarate-d6 | 66487-95-4 | 99.9% | 150.16 | 1 MG
SDP
Catalog No. 5000385000
Encompass_Preferred
Dimethyl fumarate-d6 is a deuterium-labeled Dimethyl fumarate that acts as a nuclear factor (erythroid-derived)-like 2 (Nrf2) pathway activator, leading to the upregulation of antioxidant gene expression. It is intended for research use only and can be used for various analytical purposes.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS
Catalog No. 50-003-85000 Supplier Medchemexpress LLC Supplier No. HY17363S1MG
Dimethyl fumarate-d6 is a deuterium-labeled Dimethyl fumarate that acts as a nuclear factor (erythroid-derived)-like 2 (Nrf2) pathway activator, leading to the upregulation of antioxidant gene expression. It is intended for research use only and can be used for various analytical purposes.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS