Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dimethyl fumarate-d6 | 66487-95-4 | MFCD29045610 | >95.0% | 150.16 g/mol | C6H2D6O4 | 10 MG
SDP

Catalog No. 5000294734
Encompass_Preferred

Dimethyl fumarate-d6 is a deuterium-labeled analogue of dimethyl fumarate used as a research reagent and analytical standard. It functions as an Nrf2 pathway activator and is employed in studies of antioxidant gene regulation and related signaling. The compound is provided as a solid, with molecular weight 150.16 g/mol and CAS 66487-95-4.

  • Deuterium-labeled (d6) analogue for tracer and quantitation studies.
  • Activates Nrf2 pathway; useful for oxidative stress research.
  • Isotopic purity >95% by NMR; ensures labeling integrity.
  • Chemical purity >95.0% (HPLC), suitable for analytical use.
  • Molecular weight 150.16 g/mol; formula C6H2D6O4.
  • Supplied as a solid, typically used in milligram quantities for assays.

Catalog No. 50-002-94734 Supplier Medchemexpress LLC Supplier No. HY17363S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dimethyl fumarate-d6 is a deuterium-labeled analogue of dimethyl fumarate used as a research reagent and analytical standard. It functions as an Nrf2 pathway activator and is employed in studies of antioxidant gene regulation and related signaling. The compound is provided as a solid, with molecular weight 150.16 g/mol and CAS 66487-95-4.

  • Deuterium-labeled (d6) analogue for tracer and quantitation studies.
  • Activates Nrf2 pathway; useful for oxidative stress research.
  • Isotopic purity >95% by NMR; ensures labeling integrity.
  • Chemical purity >95.0% (HPLC), suitable for analytical use.
  • Molecular weight 150.16 g/mol; formula C6H2D6O4.
  • Supplied as a solid, typically used in milligram quantities for assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.