Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Homoterephthalate 98.0+%, TCI America™
SDP

Catalog No. D52665G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D52665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D52665G Supplier TCI America Supplier No. D52665G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 52787-14-1
Molecular Formula C11H12O4
Molecular Weight (g/mol) 208.213
MDL Number MFCD07779210
InChI Key QAQYBHOZQQRJBA-UHFFFAOYSA-N
Synonym Homoterephthalic Acid Dimethyl Ester, Methyl 4-Methoxycarbonylphenylacetate, 4-Methoxycarbonylphenylacetic Acid Methyl Ester, Methyl 4-(2-Methoxy-2-oxoethyl)benzoate, 4-(2-Methoxy-2-oxoethyl)benzoic Acid Methyl Ester
PubChem CID 10536335
IUPAC Name methyl 4-(2-methoxy-2-oxoethyl)benzoate
SMILES COC(=O)CC1=CC=C(C=C1)C(=O)OC

Specifications

Color White-Yellow
Boiling Point 122°C
Quantity 5 g
Formula Weight 208.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Dimethyl Homoterephthalate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.