Learn More
Sigma Aldrich Fine Chemicals Biosciences Dimethyl isosorbide BioRen
Catalog No. 502903613
Encompass
We are committed to bringing you Greener Alternative Products which adhere to one or more of The 12 Principles of Greener Chemistry. Dimethyl isosorbide (DMI) is a biobased greener solvent and thus aligns with Use of Renewable feedstocks and Safer Solvents and Auxiliaries. Click here for more information.
Catalog No. 50-290-3613 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 9068324L
We are committed to bringing you Greener Alternative Products which adhere to one or more of The 12 Principles of Greener Chemistry. Dimethyl isosorbide (DMI) is a biobased greener solvent and thus aligns with Use of Renewable feedstocks and Safer Solvents and Auxiliaries. Click here for more information.