Learn More
TARGETMOL CHEMICALS INC DIMETHYL ITACONATE 500MG
SDP
Catalog No. 502579781
Encompass_Preferred
Also available in 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dimethyl Itaconate is a chemical reagent used in the synthesis of functionalized 2-isoxazolines used in fragment-based drug discovery. purity: 99%
Catalog No. 50-257-9781 Supplier TARGETMOL CHEMICALS INC Supplier No. T5377500MG
Also available in 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dimethyl Itaconate is a chemical reagent used in the synthesis of functionalized 2-isoxazolines used in fragment-based drug discovery. purity: 99%