Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl L-(-)-Malate 98.0+%, TCI America™
SDP

Catalog No. M134325G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 617-55-0
Molecular Formula C6H10O5
Molecular Weight (g/mol) 162.14
MDL Number MFCD00066215
InChI Key YSEKNCXYRGKTBJ-UHFFFAOYNA-N
Synonym s-dimethyl 2-hydroxysuccinate, dimethyl s---malate, dimethyl l-malate, l---malic acid dimethyl ester, 1,4-dimethyl 2s-2-hydroxybutanedioate, butanedioic acid, hydroxy-, dimethyl ester, 2s, dimethyl l---malate, pubchem6778, di-methyl s-malate, dimethyl s--malate
PubChem CID 10285815
IUPAC Name 1,4-dimethyl 2-hydroxybutanedioate
SMILES COC(=O)CC(O)C(=O)OC

Specifications

Color Yellow
Boiling Point 108°C
Quantity 25 g
Formula Weight 162.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl L-(-)-Malate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.