Learn More
Medchemexpress LLC Dimethyl malonate | 108-59-8 | MFCD00008460 | 1g
SDP
Catalog No. 5000266188
Encompass_Preferred
Dimethyl malonate is a competitive inhibitor of succinate dehydrogenase (SDH) Dimethyl malonate is able to cross the blood-brain barrier and hydrolyse to malonate Dimethyl malonate reduces neuronal apoptosis[1]
Catalog No. 50-002-66188 Supplier Medchemexpress LLC Supplier No. HYY17871G
Dimethyl malonate is a competitive inhibitor of succinate dehydrogenase (SDH) Dimethyl malonate is able to cross the blood-brain barrier and hydrolyse to malonate Dimethyl malonate reduces neuronal apoptosis[1]