Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl methoxymethylenemalonate, 98+%, Thermo Scientific™

Catalog No. AAB2380309
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2380309 10 g
AAB2380318 50 g
AAB2380330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2380309 Supplier Thermo Scientific Chemicals Supplier No. B2380309

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22398-14-7
Molecular Formula C7H10O5
Molecular Weight (g/mol) 174.152
MDL Number MFCD00038225
InChI Key RHFZTBSULNJWEI-UHFFFAOYSA-N
Synonym dimethyl methoxymethylenemalonate, dimethyl 2-methoxymethylene malonate, 1,3-dimethyl 2-methoxymethylidene propanedioate, methoxymethylenemalonic acid dimethyl ester, dimethyl methoxymethylene malonate, acmc-1cq9e, dimethylmethoxymethylenemalonate, propanedioic acid, methoxymethylene-, dimethyl ester, dimethyl methoxymethylidenemalonate
PubChem CID 90770
IUPAC Name dimethyl 2-(methoxymethylidene)propanedioate
SMILES COC=C(C(=O)OC)C(=O)OC

Specifications

Melting Point 41°C to 45°C
Boiling Point 97°C to 99°C (0.5 mmHg)
Flash Point 158°C (316°F)
Odor Ester-like
Quantity 10 g
Formula Weight 174.16
Percent Purity ≥98%
Chemical Name or Material Dimethyl methoxymethylenemalonate
Hazard Category H302-H315-H317-H319
Hazard Statement GHS H Statement
H301-H334-H315-H319-H317-H335
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P330-P333+P313-P362-P501c
EINECSNumber 244-957-4
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.