Learn More
Medchemexpress LLC Dimethyl methylene blue zinc chloride double salt | 931418-92-7 | MFCD19686959 | 99.6% | 416.04 | C18H22N3S·1/2ZnCl4 | 250mg
SDP
Catalog No. 5000269287
Encompass_Preferred
Dimethyl methylene blue zinc chloride double salt (DMMB) is a cationic dye Dimethyl methylene blue zinc chloride double salt competitively binds to the active site of the PANI-Benz-Ag nanocomposite with endotoxin releasing DMMB and increasing the fluorescence intensity Dimethyl methylene blue zinc chloride double salt can be used as a fluorescent probe for the detection of bacteria (such as E Coli) endotoxin[1]
Catalog No. 50-002-69287 Supplier Medchemexpress LLC Supplier No. HYW751238250MG
Dimethyl methylene blue zinc chloride double salt (DMMB) is a cationic dye Dimethyl methylene blue zinc chloride double salt competitively binds to the active site of the PANI-Benz-Ag nanocomposite with endotoxin releasing DMMB and increasing the fluorescence intensity Dimethyl methylene blue zinc chloride double salt can be used as a fluorescent probe for the detection of bacteria (such as E Coli) endotoxin[1]