Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dimethyl Methylphosphonate

Catalog No. 503119122 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-311-9122 5 g
D169102100G 100 g
D169102500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 50-311-9122 Supplier Sigma Aldrich Supplier No. D1691025G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dimethyl methylphosphonate is a phosphorus-containing organic compound, commonly used as a flame retardant. It is also used in the preparation of +a-ketophosphonates from esters.Used to test SXFA-coated SAW chemical sensors for organophosphorous compound detectionUsed as a chemical agent simulant for testing of protective performance of PSS membranes and functionalized sorbent membranesReactant for photocatalytic oxidationAdditive for synthesis of: Flame retardant high phosphorous containing unsaturated polyester resinUV-cured epoxy acrylate / microencapsulated phase change material

Specifications

CAS 77-78-1
Melting Point -32°C
Density 1.333 g/mL at 25°C
Boiling Point 188°C
Molecular Formula C2H6O4S
Refractive Index n20/D 1.413
Linear Formula (CH3O)2SO2
MDL Number MFCD00008416
Quantity 5 g
Synonym Dimethoxymethyl phosphine oxide; Fyrol DMMP; Methanephosphonic acid dimethyl ester; NSC 62240; Reoflam DMMP
Molecular Weight (g/mol) 126.13 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.