Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl N,N-diisopropylphosphoramidite, Thermo Scientific™

Catalog No. AAH2772306
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2772306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2772306 Supplier Thermo Scientific Chemicals Supplier No. H2772306

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122194-07-4
Molecular Formula C8H20NO2P
Molecular Weight (g/mol) 193.227
MDL Number MFCD00153505
InChI Key KXUMNSXPAYCKPR-UHFFFAOYSA-N
Synonym dimethyl n,n-diisopropylphosphoramidite, diisopropylaminodimethoxyphosphine, phosphoramidous acid,n,n-bis 1-methylethyl-, dimethyl ester, dimethoxyphosphanyl diisopropylamine, acmc-1cby2, dimethyl diisopropylphosphoramidite, dimethyl-n,n-diisopropylphosphoramidite, dimethyl n,n-diisopropylphosphoramidite gc
PubChem CID 5171659
IUPAC Name N-dimethoxyphosphanyl-N-propan-2-ylpropan-2-amine
SMILES CC(C)N(C(C)C)P(OC)OC

Specifications

Linear Formula (CH3O)2PN[CH(CH3)2]2
Quantity 5 g
Sensitivity Very Hygroscopic
Formula Weight 193.23
Chemical Name or Material Dimethyl N,N-diisopropylphosphoramidite
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.