Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dimethyl pentadecanedioate | 36575-82-3 | MFCD00673462 | 99.0% | 300.43 g/mol | C17H32O4 | 50 G
SDP

Catalog No. 5000281440
Encompass_Preferred

Dimethyl pentadecanedioate is the dimethyl ester of pentadecanedioic acid and is supplied for laboratory research use as a chemical reagent and analytical standard. It is suitable for use in organic synthesis, method development, and other analytical applications requiring a high-purity long-chain diester.

  • Purity approximately 99%.
  • Molecular weight 300.43 g/mol.
  • Molecular formula C17H32O4.
  • CAS number 36575-82-3.
  • Pack size 50 G.
  • Suitable for organic synthesis and analytical method development.

Catalog No. 50-002-81440 Supplier Medchemexpress LLC Supplier No. HYW46011950G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dimethyl pentadecanedioate is the dimethyl ester of pentadecanedioic acid and is supplied for laboratory research use as a chemical reagent and analytical standard. It is suitable for use in organic synthesis, method development, and other analytical applications requiring a high-purity long-chain diester.

  • Purity approximately 99%.
  • Molecular weight 300.43 g/mol.
  • Molecular formula C17H32O4.
  • CAS number 36575-82-3.
  • Pack size 50 G.
  • Suitable for organic synthesis and analytical method development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.