Learn More
Abcam DABCYL SE, 25MG
Catalog No. 50996199
Encompass_Preferred
MW 366.4 Da, Purity >98%. Long wavelength azo quencher - useful as a reagent to label proteins or peptides through their amino-groups. The succinimidyl ester is reactive with terminal amines or lysines of peptides and other nucleophiles.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-199 Supplier Abcam Supplier No. AB27540525MG
MW 366.4 Da, Purity >98%. Long wavelength azo quencher - useful as a reagent to label proteins or peptides through their amino-groups. The succinimidyl ester is reactive with terminal amines or lysines of peptides and other nucleophiles.
The product is subject to the following: Abcam Restricted Use Statement