Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dimethyl phosphite

Catalog No. 501796659
Encompass
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-179-6659 500 g
50-179-6658 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-179-6659 Supplier Sigma Aldrich Supplier No. D178454500G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Dimethyl phosphite may be used as an effective alternative to organic tin hydrides as radical reducing agents for organic halides, thionoesters, and isocyanides to form the corresponding hydrocarbons
  • Radical translocation/cyclization reactions of terminal alkynes to form cyclic phosphonates can also be effectively carried using dimethyl phosphite as a reagent under tin-free condition
TRUSTED_SUSTAINABILITY

Specifications

CAS 868-85-9
Density 1.2 g/mL (at 25°C (literature))
Boiling Point 170°C to 171°C (lit.)
Molecular Formula C2H7O3P
Refractive Index n20/D 1.402 (literature)
Linear Formula (CH3O)2 P(O)H
Quantity 500 g
MDL Number MFCD00044633
Molecular Weight (g/mol) 110.05
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.