Learn More
Sigma Aldrich Dimethyl phosphite
Description
- Dimethyl phosphite may be used as an effective alternative to organic tin hydrides as radical reducing agents for organic halides, thionoesters, and isocyanides to form the corresponding hydrocarbons
- Radical translocation/cyclization reactions of terminal alkynes to form cyclic phosphonates can also be effectively carried using dimethyl phosphite as a reagent under tin-free condition
Specifications
Specifications
| CAS | 868-85-9 |
| Density | 1.2 g/mL (at 25°C (literature)) |
| Boiling Point | 170°C to 171°C (lit.) |
| Molecular Formula | C2H7O3P |
| Refractive Index | n20/D 1.402 (literature) |
| Linear Formula | (CH3O)2 P(O)H |
| MDL Number | MFCD00044633 |
| Quantity | 25 g |
| Molecular Weight (g/mol) | 110.05 |
| Percent Purity | 98% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.