Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl phthalate-3,4,5,6-d4, MilliporeSigma™ Supelco™

Catalog No. 111018269 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8269 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8269 Supplier MilliporeSigma Supelco Supplier No. 3418925MG
Add to Cart
Add to Cart
Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 93951-89-4
Physical Form Neat
Linear Formula C6D4(CO2 CH3)2
MDL Number MFCD00144144
Melting Point 2°C
Boiling Point 282°C
Type Analytical Standard
Quantity 25 mg
Formula Weight 198.21
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.