Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dimethyl phthalate (Ring-d4) | 93951-89-4 | 1 MG
SDP

Catalog No. 5000418619
Encompass_Preferred

Dimethyl phthalate (Ring-d4) is the deuterium-labeled version of Dimethyl phthalate, a laboratory chemical primarily used for research purposes and in the manufacture of substances. This compound acts as an endocrine disruptor and a widespread pollutant, frequently employed as a plasticizer to enhance the flexibility of rigid polyvinylchloride (PVC) resins. It possesses a molecular formula of C10H6D4O4 and a molecular weight of 198.21. As a liquid, it exhibits a relative density of 1.214 g/cm³ and a melting/freezing point of 2°C, and is stable under recommended storage conditions.

  • Used as a research chemical and for manufacturing substances.
  • Functions as an endocrine disruptor and ubiquitous pollutant.
  • Commonly used as a plasticizer for rigid polyvinylchloride (PVC) resins.
  • Molecular formula: C10H6D4O4; Molecular weight: 198.21.
  • Liquid with a relative density of 1.214 g/cm³.
  • Melting/freezing point of 2°C.

Catalog No. 50-004-18619 Supplier Medchemexpress LLC Supplier No. HYN7106S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dimethyl phthalate (Ring-d4) is the deuterium-labeled version of Dimethyl phthalate, a laboratory chemical primarily used for research purposes and in the manufacture of substances. This compound acts as an endocrine disruptor and a widespread pollutant, frequently employed as a plasticizer to enhance the flexibility of rigid polyvinylchloride (PVC) resins. It possesses a molecular formula of C10H6D4O4 and a molecular weight of 198.21. As a liquid, it exhibits a relative density of 1.214 g/cm³ and a melting/freezing point of 2°C, and is stable under recommended storage conditions.

  • Used as a research chemical and for manufacturing substances.
  • Functions as an endocrine disruptor and ubiquitous pollutant.
  • Commonly used as a plasticizer for rigid polyvinylchloride (PVC) resins.
  • Molecular formula: C10H6D4O4; Molecular weight: 198.21.
  • Liquid with a relative density of 1.214 g/cm³.
  • Melting/freezing point of 2°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.