Learn More
Medchemexpress LLC Dimethyl phthalate (Ring-d4) | 93951-89-4 | MFCD00144144 | 10 MG
SDP
Catalog No. 5000419464
Encompass_Preferred
Dimethyl phthalate (Ring-d4) is a deuterium-labeled form of Dimethyl phthalate. Dimethyl phthalate is identified as an endocrine disruptor and a ubiquitous pollutant among phthalate esters (PAEs). It is commonly employed as a plasticizer to provide flexibility to rigid polyvinylchloride (PVC) resins.
  • Can be used as a tracer
  • Serves as an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS
  • Supports pharmacokinetic and metabolic studies as a stable heavy isotope tracer
Catalog No. 50-004-19464 Supplier Medchemexpress LLC Supplier No. HYN7106S10MG
Dimethyl phthalate (Ring-d4) is a deuterium-labeled form of Dimethyl phthalate. Dimethyl phthalate is identified as an endocrine disruptor and a ubiquitous pollutant among phthalate esters (PAEs). It is commonly employed as a plasticizer to provide flexibility to rigid polyvinylchloride (PVC) resins.
  • Can be used as a tracer
  • Serves as an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS
  • Supports pharmacokinetic and metabolic studies as a stable heavy isotope tracer