Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl pimelimidate, dihydrochloride, 98.9%, MP Biomedicals™

Catalog No. ICN15094505 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15094505 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15094505 Supplier MP Biomedicals, Inc Supplier No. 0215094505
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 58537-94-3
Molecular Formula C9H18N2O2
Molecular Weight (g/mol) 186.26
MDL Number MFCD00038942
InChI Key LRPQMNYCTSPGCX-UHFFFAOYSA-N
Synonym dimethyl pimelimidate dihydrochloride, pimelimidic acid dimethyl ester dihydrochloride, acmc-209tih, dimethyl pimelinediimidate dihydrochloride, dimethyl pimelinediimidate dihydro-chloride, dimethyl heptanebis imidate dihydrochloride, dimethyl pimelimidate dihydrochloride, powder, dimethyl pimelinediimidate paragraph signthornnieani, dimethyl pimelimidate dihydrochloride at
PubChem CID 11402688
IUPAC Name 1,7-dimethyl heptanediimidate
SMILES COC(=N)CCCCCC(=N)OC

Specifications

Color White
Assay Percent Range 98.9%
Quantity 5 g
Formula Weight 259.2
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl pimelimidate, dihydrochloride
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.