Learn More
Research Products International Corp Dimethyl-POPOP, 5 G

Supplier: Research Products International Corp 111007
- CAS Number: 3073-87-8
- Molecular Formula: C26H20N2O2
- Molecular Weight: 392.44
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.